Learn More
Waters Corp Waters Buffer Solution, pH 4, 1 Liter, NIST Traceable, Mercury-Free
Catalog No. 50465878 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This pH Buffer is directly traceable to NIST SRMs, mercury-free, guaranteed stable for at least one year after receipt, and is supplied with a full certificate of analysis.
  • Directly traceable to NIST SRMs.
  • Mercury-free.
  • Guaranteed stable for at least one year after receipt.
  • Supplied with a full certificate of analysis.
Catalog No. 50-465-878 Supplier Waters Corp Supplier No. 129
This pH Buffer is directly traceable to NIST SRMs, mercury-free, guaranteed stable for at least one year after receipt, and is supplied with a full certificate of analysis.
  • Directly traceable to NIST SRMs.
  • Mercury-free.
  • Guaranteed stable for at least one year after receipt.
  • Supplied with a full certificate of analysis.