Learn More
Inorganic Ventures pH 4 CALIBRATION STD 4L
SDP
Catalog No. 507527001
Encompass_Preferred
Inorganic Ventures' pH 4 Standard is a 4L certified reference material set in a water matrix for stability. This standard is Traceable to NIST and manufactured in accordance with our stringent Quality Assurance guidelines. See Certificate of Analysis for expiration.
ATTENTION: Inorganic Ventures is no longer supplying paper copies of Certificate of Analyses(CoA) and Safety Data Sheets (SDS). Please use this link to download your copies: https://www.inorganicventures.com/coa-sds-search
Catalog No. 50-752-7001 Supplier Inorganic Ventures Supplier No. PH44L
Inorganic Ventures' pH 4 Standard is a 4L certified reference material set in a water matrix for stability. This standard is Traceable to NIST and manufactured in accordance with our stringent Quality Assurance guidelines. See Certificate of Analysis for expiration.
ATTENTION: Inorganic Ventures is no longer supplying paper copies of Certificate of Analyses(CoA) and Safety Data Sheets (SDS). Please use this link to download your copies: https://www.inorganicventures.com/coa-sds-search