Learn More
Cole-Parmer Oakton pH 5+ Handheld Meter with ATC; Order pH Probe Separately
Catalog No. NC0416489 Shop All Cole-Parmer Products
Encompass_Preferred
These meters are perfect for fast, accurate results. Automatic temperature compensation (ATC) ensures accuracy from 0 to 80°C. Meters include a protective rubber boot that also acts as a meter stand.
Catalog No. NC0416489 Supplier Cole-Parmer Supplier No. 3561350
These meters are perfect for fast, accurate results. Automatic temperature compensation (ATC) ensures accuracy from 0 to 80°C. Meters include a protective rubber boot that also acts as a meter stand.