Learn More
Inorganic Ventures pH 5 CALIBRATION STD 1L (Inorganic Ventures)
SDP
Catalog No. 507526998
Encompass_Preferred
Inorganic Ventures' pH 5 Standard is a 1L certified reference material set in a water matrix for stability. This standard is Traceable to NIST and manufactured in accordance with our stringent Quality Assurance guidelines. One year Expiration Date.
ATTENTION: Inorganic Ventures is no longer supplying paper copies of Certificate of Analyses(CoA) and Safety Data Sheets (SDS). Please use this link to download your copies: https://www.inorganicventures.com/coa-sds-search
This item is not returnable. View return policy
Catalog No. 50-752-6998 Supplier Inorganic Ventures Supplier No. PH51L
Inorganic Ventures' pH 5 Standard is a 1L certified reference material set in a water matrix for stability. This standard is Traceable to NIST and manufactured in accordance with our stringent Quality Assurance guidelines. One year Expiration Date.
ATTENTION: Inorganic Ventures is no longer supplying paper copies of Certificate of Analyses(CoA) and Safety Data Sheets (SDS). Please use this link to download your copies: https://www.inorganicventures.com/coa-sds-search