Learn More
Cole-Parmer Oakton PH 550 Benchtop pH Meter Kit
Catalog No. 501961325 Shop All Cole-Parmer Products
Encompass_Preferred
The PH 550 benchtop pH meter features a microprocessor chip which provides automatic temperature compensation, data storage, parameter setup, and Max/Min reading display. The meter offers a pH range from 0 to 14 and a three-point automatic calibration, with the ability to recognize standard buffer solutions (USA or NIST series). The advanced digital processing technology improves meter response time and measuring accuracy. A stable reading display icon appears when the measuring value is stable. The 3-in-1 combination pH electrode measures pH and temperature simultaneously. The compact meter includes a detachable electrode holder and three bottles of pH buffer solution. It has a dustproof and spillproof rating of IP54. Connectors are protected by silicone seal caps.
Catalog No. 50-196-1325 Supplier Cole-Parmer Supplier No. 3541930
The PH 550 benchtop pH meter features a microprocessor chip which provides automatic temperature compensation, data storage, parameter setup, and Max/Min reading display. The meter offers a pH range from 0 to 14 and a three-point automatic calibration, with the ability to recognize standard buffer solutions (USA or NIST series). The advanced digital processing technology improves meter response time and measuring accuracy. A stable reading display icon appears when the measuring value is stable. The 3-in-1 combination pH electrode measures pH and temperature simultaneously. The compact meter includes a detachable electrode holder and three bottles of pH buffer solution. It has a dustproof and spillproof rating of IP54. Connectors are protected by silicone seal caps.