Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bio X Cell InVivoPure pH 6.5, Sterile, 50 mL

Catalog No. NC1957862
Encompass_Preferred

InVivoPure™ dilution buffers are specifically formulated and tested to satisfy the stringent requirements for in vivo applications. They are extremely low in endotoxin, have been screened for murine pathogens, tested in animal models for toxicity, and are formulated with respect to buffer composition and pH to satisfy the requirements of Bio X Cell's antibodies.

  • Endotoxin: <0.5 EU/mL (<0.0005 EU/μL)
  • Sterility: 0.2 μM filtered
  • Negative for murine pathogens (e.g., Norovirus, Parvovirus)
  • Nontoxic and nonantigenic in animal models
  • Concentration: 1X
  • Volume: 50 mL
  • Composition: 16 mM Na2HPO4, 23 mM NaH2PO4, 136 mM NaCl; free of Ca, Mg, phenol red, and azide
  • Storage: 4°C; do not freeze; keep sterile, open in biological safety cabinet

Catalog No. NC1957862 Supplier Bio X Cell Supplier No. IPT065R050ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

InVivoPure™ dilution buffers are specifically formulated and tested to satisfy the stringent requirements for in vivo applications. They are extremely low in endotoxin, have been screened for murine pathogens, tested in animal models for toxicity, and are formulated with respect to buffer composition and pH to satisfy the requirements of Bio X Cell's antibodies.

  • Endotoxin: <0.5 EU/mL (<0.0005 EU/μL)
  • Sterility: 0.2 μM filtered
  • Negative for murine pathogens (e.g., Norovirus, Parvovirus)
  • Nontoxic and nonantigenic in animal models
  • Concentration: 1X
  • Volume: 50 mL
  • Composition: 16 mM Na2HPO4, 23 mM NaH2PO4, 136 mM NaCl; free of Ca, Mg, phenol red, and azide
  • Storage: 4°C; do not freeze; keep sterile, open in biological safety cabinet

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.