Learn More
Boston Bioproducts Inc Tris-HCl Buffer (1 M, pH 6.8) - 1L
Catalog No. NC1299109
Encompass
The Tris-HCl Buffer is a widely used bufering reagent in molecular biology and biochemistry, acting to maintain a stable pH, and preserve the integrity of target macromolecules. The buffering capacity of Tris-HCl is effective in the pH range of 7.0 to 9.0, making it ideal for many biological applications like cell culture, molecular biology, DNA/RNA extraction, electrophoresis, enzyme assays and immunohistostaining.
Catalog No. NC1299109 Supplier Boston Bioproducts Inc Supplier No. BBT681L
The Tris-HCl Buffer is a widely used bufering reagent in molecular biology and biochemistry, acting to maintain a stable pH, and preserve the integrity of target macromolecules. The buffering capacity of Tris-HCl is effective in the pH range of 7.0 to 9.0, making it ideal for many biological applications like cell culture, molecular biology, DNA/RNA extraction, electrophoresis, enzyme assays and immunohistostaining.