Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

METTLER TOLEDO 3P INDUSTRIAL Technical buffer pH 7.00, 6 x 250mL

Catalog No. NC1820240
Encompass_Preferred

pH 7.00, 6 Bottles x 250 mL, Shelf Life: 24 months

Catalog No. NC1820240 Supplier METTLER TOLEDO 3P INDUSTRIAL Supplier No. 51350020
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

pH 7.00, 6 Bottles x 250 mL, Shelf Life: 24 months

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.