Learn More
Andwin Scientific PH 7.0 YELLOW REAGENT SOL
SDP
Catalog No. NC9888475
Encompass_Preferred
The BDH pH reference standard buffer is a clear, yellow, odorless, liquid solution. This contains hydrochloric acid, potassium chloride, and deionized water. It is classified to have a 7.00 pH at 25.0 �C. This product is packaged and sold in a 500 mL poly bottle.
Catalog No. NC9888475 Supplier Andwin Scientific Supplier No. BDH5046
The BDH pH reference standard buffer is a clear, yellow, odorless, liquid solution. This contains hydrochloric acid, potassium chloride, and deionized water. It is classified to have a 7.00 pH at 25.0 �C. This product is packaged and sold in a 500 mL poly bottle.