Learn More
Cole-Parmer Thermo Scientific 910107 pH Buffer Solution, 7.00, Yellow; 475 mL Bottle
Catalog No. NC0310118 Shop All Cole-Parmer Products
Encompass_Preferred
Thermo Scientific Orion pH buffers are available in 475-mL (pint) bottles and in portable 5 x 60 mL convenience packs. Portable pH buffers are easier to carry than larger bottles, minimize wasted buffer, and provide more volume than pouches. Each buffer is color-coded for easy identification and packaged with buffer/temperature tables for accurate calibrations. Buffers are standardized against NIST-traceable pH references for superior accuracy.
Catalog No. NC0310118 Supplier Cole-Parmer Supplier No. 5535046
Thermo Scientific Orion pH buffers are available in 475-mL (pint) bottles and in portable 5 x 60 mL convenience packs. Portable pH buffers are easier to carry than larger bottles, minimize wasted buffer, and provide more volume than pouches. Each buffer is color-coded for easy identification and packaged with buffer/temperature tables for accurate calibrations. Buffers are standardized against NIST-traceable pH references for superior accuracy.