Learn More
Medchemexpress LLC PH-797804 | 586379-66-0 | 98.9% | 477.30 g·mol⁻¹ | C22H19BrF2N2O3 | 200 MG
SDP
Catalog No. 5000242502
Encompass_Preferred
PH-797804 is an ATP-competitive, selective inhibitor of p38 mitogen-activated protein kinases (p38α/p38β) used in research to probe p38-mediated signaling and inflammatory responses. It shows nanomolar biochemical potency and demonstrable cellular activity that reduces LPS-induced cytokine production.
  • Selective ATP-competitive p38α/p38β inhibitor.
  • Nanomolar biochemical potency (p38α IC50 26 nM; Ki 5.8 nM).
  • Demonstrated cellular activity reducing LPS-induced TNF-α and IL-1β release.
  • Does not inhibit JNK2, supporting target selectivity.
  • High supplied purity and solid white to off-white appearance.
  • Suitable for biochemical and cellular assay applications.
Catalog No. 50-002-42502 Supplier Medchemexpress LLC Supplier No. HY10403200MG
PH-797804 is an ATP-competitive, selective inhibitor of p38 mitogen-activated protein kinases (p38α/p38β) used in research to probe p38-mediated signaling and inflammatory responses. It shows nanomolar biochemical potency and demonstrable cellular activity that reduces LPS-induced cytokine production.
  • Selective ATP-competitive p38α/p38β inhibitor.
  • Nanomolar biochemical potency (p38α IC50 26 nM; Ki 5.8 nM).
  • Demonstrated cellular activity reducing LPS-induced TNF-α and IL-1β release.
  • Does not inhibit JNK2, supporting target selectivity.
  • High supplied purity and solid white to off-white appearance.
  • Suitable for biochemical and cellular assay applications.