Learn More
ERA WATERS CORP PH
SDP
Catalog No. NC9757329
Encompass
One 15 mL screw-cap vial yields up to 1 liter after dilution. Use with nephelometric methods.
Catalog No. NC9757329 Supplier ERA WATERS CORP Supplier No. 552
One 15 mL screw-cap vial yields up to 1 liter after dilution. Use with nephelometric methods.