Learn More
Chemglass Life Sciences pH and DO Probe Rack
SDP
Catalog No. NC1797074
Encompass
Freestanding rack can hold 36 probes (pH or DO) up to 235mm long.Rack is constructed of polypropylene with stainless steel uprights.Footprint of rack 10. 5" Length x 4.5" width.pH and DO probes must be purchased seperately.
Catalog No. NC1797074 Supplier Chemglass Life Sciences Supplier No. CLS1455R1
Freestanding rack can hold 36 probes (pH or DO) up to 235mm long.Rack is constructed of polypropylene with stainless steel uprights.Footprint of rack 10. 5" Length x 4.5" width.pH and DO probes must be purchased seperately.