Learn More
Chemglass Life Sciences Complete Baffles for use in 30L-50L (Short) Reactors: Overall Baffle Length: 34", Effective Baffle Length: 14", Baffle Width x Depth (mm): 20 x 30, Lower Compression TFE Fitting: 3/4" to 12mm, Adapter Joint Size: 45/50
SDP
Catalog No. CG1872B10
Encompass_Preferred
Features: Available in Square or Rectangular Shapes, with Open BottomPTFE Joint is Offset Permitting Baffle to be Adjusted up to 1" Towards the Center of the ReactorBaffle Can Be Vertically Adjusted Inside ReactorHollow Baffles Allow pH Electrodes to be Inserted Into the Reaction¿Complete Baffles Consist of: Glass Baffle, Glass Fillted PTFE Adapter, and Lower PTFE Compression Fitting. glass baffle system for process reactors from 10L to 50L. Designed to increase agitation in the reactor and mimic baffles found in larger glass lined reactors. All baffles have 3/4" precision ground top and lower tubing for use with compression fittings and top jointed adapter. PLEASE NOTE: SEE CG-1971 FOR THERMOCOUPLE BAFFLES AND CG-1872 FOR pH ELECTRODES. ¿Designed and developed by John Korzun, Bristol Myers Squibb,Brunswick, NJ
Catalog No. CG1872B10 Supplier Chemglass Life Sciences Supplier No. CG1872B10
Features: Available in Square or Rectangular Shapes, with Open BottomPTFE Joint is Offset Permitting Baffle to be Adjusted up to 1" Towards the Center of the ReactorBaffle Can Be Vertically Adjusted Inside ReactorHollow Baffles Allow pH Electrodes to be Inserted Into the Reaction¿Complete Baffles Consist of: Glass Baffle, Glass Fillted PTFE Adapter, and Lower PTFE Compression Fitting. glass baffle system for process reactors from 10L to 50L. Designed to increase agitation in the reactor and mimic baffles found in larger glass lined reactors. All baffles have 3/4" precision ground top and lower tubing for use with compression fittings and top jointed adapter. PLEASE NOTE: SEE CG-1971 FOR THERMOCOUPLE BAFFLES AND CG-1872 FOR pH ELECTRODES. ¿Designed and developed by John Korzun, Bristol Myers Squibb,Brunswick, NJ