Learn More
Eurofins Discoverx PathHunter Bioassay Kit ED Detection Kit, 10 (200 dp, 96-well). (93-1043E)
Catalog No. 502001914
Encompass
The PathHunter Bioassay ED Detection Kit is a detection kit intended for use with Eurofins DiscoverX's PathHunter Bioassay cells overexpressing ED (ProLabel/PK) but not containing any EA. This kit contains lysis buffer, exogenous EA, and EFC substrate to allow for detection of levels of ED in the assayed cells. This kit contains all components required to assay 2 plates (96-well). For detailed product information, User manual and COA, visit www.discoverx.com/products/reagents/detection-kit-93-1043e.
Catalog No. 50-200-1914 Supplier Eurofins Discoverx Supplier No. 931043E
The PathHunter Bioassay ED Detection Kit is a detection kit intended for use with Eurofins DiscoverX's PathHunter Bioassay cells overexpressing ED (ProLabel/PK) but not containing any EA. This kit contains lysis buffer, exogenous EA, and EFC substrate to allow for detection of levels of ED in the assayed cells. This kit contains all components required to assay 2 plates (96-well). For detailed product information, User manual and COA, visit www.discoverx.com/products/reagents/detection-kit-93-1043e.