Learn More
Eurofins Discoverx PathHunter Bioassay Kit ED Detection Kit, 50 (1,000 dp, 96-well). (93-1043)
Catalog No. 502001913
Encompass
The PathHunter Bioassay ED Detection Kit is a detection kit intended for use with Eurofins DiscoverX's PathHunter Bioassay cells overexpressing ED (ProLabel/PK) but not containing any EA. This kit contains lysis buffer, exogenous EA, and EFC substrate to allow for detection of levels of ED in the assayed cells. This kit contains all components required to assay 10 plates (96-well). For detailed product information, User manual and COA, visit www.discoverx.com/products/reagents/detection-kit-93-1043.
Catalog No. 50-200-1913 Supplier Eurofins Discoverx Supplier No. 931043
The PathHunter Bioassay ED Detection Kit is a detection kit intended for use with Eurofins DiscoverX's PathHunter Bioassay cells overexpressing ED (ProLabel/PK) but not containing any EA. This kit contains lysis buffer, exogenous EA, and EFC substrate to allow for detection of levels of ED in the assayed cells. This kit contains all components required to assay 10 plates (96-well). For detailed product information, User manual and COA, visit www.discoverx.com/products/reagents/detection-kit-93-1043.