Learn More
Andwin Scientific PH BUFFER 12.45
SDP
Catalog No. NC9708660
Encompass_Preferred
The BDH pH reference standard buffer is a clear, colorless, odorless, liquid solution. This contains calcium hydroxide and deionized water. It is classified to have a 12.45 pH at 25.0 C. This product is packaged and sold in a 500 mL poly bottle.
Catalog No. NC9708660 Supplier Andwin Scientific Supplier No. BDH5092500ML
The BDH pH reference standard buffer is a clear, colorless, odorless, liquid solution. This contains calcium hydroxide and deionized water. It is classified to have a 12.45 pH at 25.0 C. This product is packaged and sold in a 500 mL poly bottle.