Learn More
Cole-Parmer Oakton Buffer Solution, pH 4.01; 500 mL
Supplier:  Cole-Parmer 0065400
Encompass_Preferred
Ideal for laboratory and field analysis. All Oakton pH buffers are traceable to NIST standard reference materials. Buffer solution lets you calibrate pH instruments to ensure accuracy. If unopened, shelf life is 2 years. NIST/USA buffer value of pH 4.01.
Catalog No. NC0306507