Learn More
Metrohm USA PH BUFFER 4 7 9 10X30MLPH BUF
SDP
Supplier:  Metrohm USA 62307230
Encompass
Mixed buffer solutions pH 4.00/7.00/9.00 (25 �C) in single use sachets, colourless, box of 3 x 10 x 30 mL
Catalog No. NC0626524