Learn More
Cole-Parmer Metrepak 272,PH4 pH Calibration Buffer Capsules, 4.0, orange, 10/vial
Catalog No. NC9018264 Shop All Cole-Parmer Products
Encompass_Preferred
Prepare fresh buffer solutions with these handy pH buffer capsules. Empty the contents of one capsule into 100 mL of distilled water and stir. Buffer solutions are accurate to ±0.02 pH. Each vial contains ten capsules.
Catalog No. NC9018264 Supplier Cole-Parmer Supplier No. 0597712
Prepare fresh buffer solutions with these handy pH buffer capsules. Empty the contents of one capsule into 100 mL of distilled water and stir. Buffer solutions are accurate to ±0.02 pH. Each vial contains ten capsules.