Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

VeriSpec™ pH Buffer, Certified Reference Material
SDP

Catalog No. 01100272 Shop All Ricca Chemical Company Products
Change view
Click to view available options
Quantity:
500 mL
1 L
Color:
Blue
Colorless
Red
Yellow
pH:
1.00 ±0.01
1.68 ±0.01
10.00 ±0.01
11.00 ±0.01
12.00 ±0.01
12.45 ±0.01
13.00 ±0.01
2.00 ±0.01
3.00 ±0.01
4.00 ±0.01
5.00 ±0.01
6.00 ±0.01
6.86 ±0.01
7.00 ±0.01
8.00 ±0.01
9.00 ±0.01
31 product options available for selection
Product selection table with 31 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Color pH
01-100-272 500 mL Red 4.00 ±0.01
01-185-028 500 mL Colorless 1.00 ±0.01
01-185-027 500 mL Colorless 1.68 ±0.01
01-100-270 500 mL Colorless 2.00 ±0.01
01-185-026 500 mL Colorless 3.00 ±0.01
01-185-025 500 mL Colorless 4.00 ±0.01
01-185-024 500 mL Colorless 5.00 ±0.01
01-185-023 500 mL Colorless 6.00 ±0.01
01-185-022 500 mL Colorless 6.86 ±0.01
01-185-021 500 mL Colorless 7.00 ±0.01
01-100-274 500 mL Yellow 7.00 ±0.01
01-185-020 500 mL Colorless 8.00 ±0.01
01-185-019 500 mL Colorless 9.00 ±0.01
01-185-018 500 mL Colorless 10.00 ±0.01
01-100-276 500 mL Blue 10.00 ±0.01
01-185-017 500 mL Colorless 11.00 ±0.01
01-185-016 500 mL Colorless 12.00 ±0.01
01-100-277 500 mL Colorless 12.45 ±0.01
01-185-015 500 mL Colorless 13.00 ±0.01
RV1495001A 1 L Colorless 3.00 ±0.01
RV1500001A 1 L Colorless 4.00 ±0.01
01-100-271 1 L Red 4.00 ±0.01
RV1505001A 1 L Colorless 5.00 ±0.01
RV1510001A 1 L Colorless 6.00 ±0.01
RV1540001A 1 L Colorless 6.86 ±0.01
RV1550001A 1 L Colorless 7.00 ±0.01
01-100-273 1 L Yellow 7.00 ±0.01
RV1580001A 1 L Colorless 8.00 ±0.01
RV1590001A 1 L Colorless 9.00 ±0.01
RV1600001A 1 L Colorless 10.00 ±0.01
01-100-275 1 L Blue 10.00 ±0.01
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 31 options available.
31 options
This item is not returnable. View return policy
Catalog No. 01-100-272 Supplier VeriSpec™ Supplier No. RV150100500A
Add to Cart
Add to Cart
This item is not returnable. View return policy

ISO 17025/17034 pH Buffer Certified Reference Materials provide the highest level of quality assurance. A certificate of analysis providing the specified property value, uncertainty, and traceability is available with every bottle.

  • No formaldehyde or mercury compounds as preservatives
  • No chromium compounds as colorants
  • Manufactured and tested in ISO 17025/17034 accredited facilities
  • pH 4, 7, and 10 are color coded for easy identification
  • Certified traceable to NIST

Specifications

Chemical Name or Material VeriSpec™ Certified Reference Material, Buffer pH 4.00 ±0.01 (at 25°C)
CAS Proprietary, 7732-18-5, 877-24-7
Color Red
Certificate of Analysis Yes
Concentration 1X
For Use With (Application) Calibration
For Use With (Equipment) pH Meter and Electrode
pH 4.00 ±0.01
Physical Form Liquid
Product Line VeriSpec
Quantity 500 mL
Solution Type pH Buffer
Formulation Aqueous Solution
Packaging Natural Poly
Traceability to NIST NIST Traceable
Grade Certified
Show More Show Less

ISO 17025/ISO 17034

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.