Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

pH Buffer Pack
SCI_ED

Catalog No. s02641 Shop All Fisherbrand Products
Change view
Click to view available options
Quantity:
4 x 500 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S02641 4 x 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S02641
Only null left
Add to Cart
Edge
Add to Cart

Includes

pH Buffer 4: 500mL pH Buffer 7: 500mL pH Buffer 10: 500mL Electrode Storage Solution: 500mL

Specifications

Chemical Name or Material pH Buffer Solutions
Quantity 4 x 500 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.