Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

pH Buffer Sachets, METTLER TOLEDO™
GSA_VAGREENER_CHOICE

Catalog No. 01911291 Shop All METTLER TOLEDO Products
Change view
Click to view available options
Quantity:
30 x 20 mL
pH:
10.01
4, 7, 10
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity pH
01-911-291 30 x 20 mL 4, 7, 10
01-911-289 30 x 20 mL 10.01
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 01-911-291 Supplier METTLER TOLEDO Supplier No. 51302080
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Buffer solutions in sealed sachets for easy and precise calibration of pH measurement equipment.

  • Traceable to NIST primary standards

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

TRUSTED_SUSTAINABILITY

Specifications

Color Multicolored
For Use With (Application) Calibration
For Use With (Equipment) pH Meter
pH 4, 7, 10
Physical Form Powder or Solid
Quantity 30 x 20 mL
Solution Type pH Buffer
Packaging Satchets
Grade Technical
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.