Learn More
Description
Used to maintain a standard pH level
Available in various sizes, from pH 2 to 10
Specifications
Specifications
| For Use With (Application) | pH Measurement |
| For Use With (Equipment) | pH Meter and Electrode |
| pH | 10 |
| Physical Form | Liquid |
| Quantity | 1 L |
| Solution Type | Buffer |
| Grade | Reagent |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
