Learn More
Description
- Traceable to NIST standard reference materials
- Convenient wide neck round bottle can be used as a calibration vessel and can be easily refilled
- Fits inside of Lovibond SD3XX pH meter cases
Specifications
Specifications
| For Use With (Application) | Calibration and Maintenance |
| For Use With (Equipment) | pH Meter and Electrode |
| pH | 4, 7, 10 |
| Physical Form | Liquid |
| Quantity | 90 mL |
| Solution Type | pH Buffer |
| Grade | Reagent |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.