Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Innovating Science™ pH 10 Buffer Solution
SCI_ED

Catalog No. S06494 Shop All Aldon Products
Change view
Click to view available options
Quantity:
500 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S06494 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S06494 Supplier Innovating Science™ Supplier No. ADBB0410500ML
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This pH 10 buffer solution is ideal for use in performing EDTA titrations.

500 mL Colorless Solution

Specifications

Chemical Name or Material Buffer Solutions
Concentration 0.05 M
For Use With (Application) Water Analysis
For Use With (Equipment) pH Probe
pH 10.00
Physical Form Liquid
Quantity 500 mL
Solution Type Buffer
Molecular Weight (g/mol) Mixture
Percent Purity 0.20%
Grade Reagent
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.