Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisher Science Education™ pH Buffer Solution
SCI_ED

Catalog No. S25849B Shop All Fisherbrand Products
Change view
Click to view available options
Quantity:
500 mL
Packaging:
pH 10.0: Blue
pH 4.0: Red
pH 7.0: Yellow
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
S25849B 500 mL pH 7.0: Yellow
S25849 500 mL pH 10.0: Blue
S25849A 500 mL pH 4.0: Red
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. S25849B
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Moderate Hazard

This product is for educational use - no C of A is available.

Specifications

Chemical Name or Material pH Buffer Solutions (pH 7.00)
CAS 1310-73-2, 7778-77-0, 110-44-1, 7732-18-5
Color Yellow
Density 1.000
Physical Form Liquid
Quantity 500 mL
Packaging pH 7.0: Yellow
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.