Learn More
Research Products International Corp Traceable pH Buffer Standards, 7.416 pH Value, 16oz
SDP
Catalog No. 501367792
Encompass
Traceable pH Buffers provide the most precise accuracy available, ± 0.010 pH @ 25°C. These buffers have a Certified Reference Materials Classification and meet all Federal, State and local requirements to provide exact calibration results with all pH meters. Each buffer is supplied with an individual and serialized Traceable Certificate of Calibration. Available in economical 16 ounce bottle featuring exclusive triple layer packaging that ensures a two year shelf life.
Catalog No. 50-136-7792 Supplier Research Products International Corp Supplier No. 844286
Traceable pH Buffers provide the most precise accuracy available, ± 0.010 pH @ 25°C. These buffers have a Certified Reference Materials Classification and meet all Federal, State and local requirements to provide exact calibration results with all pH meters. Each buffer is supplied with an individual and serialized Traceable Certificate of Calibration. Available in economical 16 ounce bottle featuring exclusive triple layer packaging that ensures a two year shelf life.