Learn More
Bellco Glass, Inc. pH Cable,3' Mettler
SDP
Catalog No. 501946287
Encompass_Preferred
Cable has a k9 probe connection and bnc controller connection.Size: 3"
Catalog No. 50-194-6287 Supplier Bellco Glass, Inc. Supplier No. 780321540
Cable has a k9 probe connection and bnc controller connection.Size: 3"