Learn More
Med Vet International Calcium Gluconate 23% 500mL
Catalog No. 502232638
Encompass_Preferred
Sterile, preservative-free solution for use as an aid in the treatment of parturient paresis (milk fever) in cattle. Administer intravenously or intraperitoneally. Each 100 mL contains: Calcium gluconate (provides 2.14 grams calcium), 23 grams Water for injection, q.s. Boric acid as a solubilizing agent
Catalog No. 50-223-2638 Supplier Med Vet International Supplier No. PHCALCIUMGLU
Sterile, preservative-free solution for use as an aid in the treatment of parturient paresis (milk fever) in cattle. Administer intravenously or intraperitoneally. Each 100 mL contains: Calcium gluconate (provides 2.14 grams calcium), 23 grams Water for injection, q.s. Boric acid as a solubilizing agent