Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SI Analytics™ pH Combination Electrodes

Catalog No. 15177882 Shop All Yellow Springs Instrument (YSI) Products
Change view
Click to view available options
Model:
N 61
N 62
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Model
15-177-882 N 62
15-177-881 N 61
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 15-177-882 Supplier SI Analytics™ Supplier No. 285100034
Add to Cart
Add to Cart

With Plug Head and Fixed Cable

  • For Calculating Acidity and Alkalinity with Excellence and Consistency
TRUSTED_SUSTAINABILITY

Specifications

Connector Types Plug Head
Junction Platinum Diaphragm
Material Glass
pH Range 0 to 14
Filling Solution Type 3 M KCl
For Use With (Application) Calculating Acidity and Alkalinity with Excellence and Consistency
Internal Reference Silamid™ Reference System
Membrane Sphere Membrane
Model N 62
Temperature (Metric) Range -5°C to +100°C
Length (Metric) 120 mm
Length (Metric) Cable 1 m (Fixed)
Diameter (Metric) 12 mm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.