Learn More
Cole-Parmer Oakton PC700 Benchtop Meter with Probes and NIST-Traceable Calibration
Catalog No. 501961307 Shop All Cole-Parmer Products
Encompass_Preferred
The PC700 meter offers a new compact design that is over 40% smaller than other benchtop meters. The large display is easy to read even at a distance. Dual-display allows you to read both a measured parameter and temperature at a glance. The meter features pH auto-buffer recognition for both USA and NIST buffer sets (1.68, 4.01, 7.00, 10.01 and 12.45 or 1.68, 4.01. 6.18. 9.86, and 12.45), automatic or manual temperature compensation, temperature selectable in either °F or °C, display Hold, and water-resistant membrane keypad. Previous calibration points, electrode slope and offset, and conductivity/TDS cell constants (0.1, 1.0 and 10) are stored in memory for quick and easy retrieval. You can also store up to 100 data sets in the meter's memory for retrieval later. The convenient pull-out quick reference card keeps keypad functions at your fingertips.
Catalog No. 50-196-1307 Supplier Cole-Parmer Supplier No. 3541301
The PC700 meter offers a new compact design that is over 40% smaller than other benchtop meters. The large display is easy to read even at a distance. Dual-display allows you to read both a measured parameter and temperature at a glance. The meter features pH auto-buffer recognition for both USA and NIST buffer sets (1.68, 4.01, 7.00, 10.01 and 12.45 or 1.68, 4.01. 6.18. 9.86, and 12.45), automatic or manual temperature compensation, temperature selectable in either °F or °C, display Hold, and water-resistant membrane keypad. Previous calibration points, electrode slope and offset, and conductivity/TDS cell constants (0.1, 1.0 and 10) are stored in memory for quick and easy retrieval. You can also store up to 100 data sets in the meter's memory for retrieval later. The convenient pull-out quick reference card keeps keypad functions at your fingertips.