Learn More
New England Biolabs, Inc. Ph.D.-12 Phage Display Peptide Library Kit v2 - 1 set
SDP
Catalog No. 502269738
Encompass
The Ph.D.-12 Phage Display Peptide Library Kit v2 is based on a combinatorial library of random dodecapeptides fused to the N-terminus of a minor coat protein (pIII) of M13 phage. The peptide is followed by a short spacer (Gly-Gly-Gly) which directly precedes the wild-type pIII sequence. The library consists of about 1E09 electroporated sequences amplified once to yield approximately 100 copies of each sequence in 10 ul of the supplied phage. The Ph.D.-12 Phage Display Peptide Library Kit v2 contains the Ph.D.-12 Phage Display Peptide Library a DYKDDDDK Mouse monoclonal antibody and Protein G Magnetic Beads for a panning control experiment and enough -96gIII sequencing primer for greater than 50 sequencing reactions.
This item is not returnable. View return policy
Catalog No. 50-226-9738 Supplier New England Biolabs, Inc. Supplier No. E8210S
The Ph.D.-12 Phage Display Peptide Library Kit v2 is based on a combinatorial library of random dodecapeptides fused to the N-terminus of a minor coat protein (pIII) of M13 phage. The peptide is followed by a short spacer (Gly-Gly-Gly) which directly precedes the wild-type pIII sequence. The library consists of about 1E09 electroporated sequences amplified once to yield approximately 100 copies of each sequence in 10 ul of the supplied phage. The Ph.D.-12 Phage Display Peptide Library Kit v2 contains the Ph.D.-12 Phage Display Peptide Library a DYKDDDDK Mouse monoclonal antibody and Protein G Magnetic Beads for a panning control experiment and enough -96gIII sequencing primer for greater than 50 sequencing reactions.