Learn More
New England Biolabs, Inc. Ph.D.™-C7C Phage Display Peptide Library Kit v2 - 1 set
SDP
Catalog No. 502336254
Encompass
The Ph.D.-C7C Phage Display Peptide Library Kit v2 is based on a combinatorial library of random disulfide looped peptides fused to the N-terminus of a minor coat protein (pIII) of M13 phage. The library is displayed in the form AC-X7-C-GGG, which directly precedes the wild-type pIII sequence. Under neutral conditions disulfide loop formation is favored. The library consists of ~1E09 electroporated sequences amplified once to yield approximately 100 copies of each sequence in 10 μl of the supplied phage. The Ph.D.-C7C Phage Display Peptide Library Kit v2 contains the Ph.D.-C7C Phage Display Peptide Library, a DYKDDDDK Mouse monoclonal antibody and Protein G Magnetic Beads for a panning control experiment, and enough -96gIII sequencing primer for >50 sequencing reactions.
Catalog No. 50-233-6254 Supplier New England Biolabs, Inc. Supplier No. E8212S
The Ph.D.-C7C Phage Display Peptide Library Kit v2 is based on a combinatorial library of random disulfide looped peptides fused to the N-terminus of a minor coat protein (pIII) of M13 phage. The library is displayed in the form AC-X7-C-GGG, which directly precedes the wild-type pIII sequence. Under neutral conditions disulfide loop formation is favored. The library consists of ~1E09 electroporated sequences amplified once to yield approximately 100 copies of each sequence in 10 μl of the supplied phage. The Ph.D.-C7C Phage Display Peptide Library Kit v2 contains the Ph.D.-C7C Phage Display Peptide Library, a DYKDDDDK Mouse monoclonal antibody and Protein G Magnetic Beads for a panning control experiment, and enough -96gIII sequencing primer for >50 sequencing reactions.