Learn More
METTLER TOLEDO pH Sensor InPro 3250SG 120/PT100
Supplier:  METTLER TOLEDO 52002558
Encompass
The InPro 3250SG 120/PT100 is a pre-pressurized, liquid filled, low maintenance pH/ORP sensor, combined with a PT100 temperature sensor, for in-line measurement in demanding applications. The 120 mm version is optimal for short installation lengths.
Catalog No. NC9659059