Learn More
YSI 3P LifeScience PH ELECTROD SENTIX 81 DIN
Catalog No. 50465214
Encompass_Preferred
Glass pH combination electrode with built-in temperature sensor, waterproof DIN plug and banana plug, length 120 mm, 3.3 ft (1 m) cable.
Catalog No. 50-465-214 Supplier YSI 3P LifeScience Supplier No. 103642
Glass pH combination electrode with built-in temperature sensor, waterproof DIN plug and banana plug, length 120 mm, 3.3 ft (1 m) cable.