Learn More
METTLER TOLEDO PH ELECTRODE 405-DPAS-SC-K8S
Supplier:  METTLER TOLEDO 59903230
Encompass
pH Electrode, Pressure resistant up to 2.5 bar, sterilization at 140deg.C is possible. No diaphragm clogging in sulfide-bearing media due to proprietary silver-ion trap
Catalog No. NC9039288