Learn More
Cole-Parmer Oakton pH Electrode, Epoxy, Double Junction, 9-mm OD
Supplier:  Cole-Parmer 3580403
Encompass_Preferred
Replace electrodes when they break, no longer hold calibration, cannot be rehydrated, or are not compatible with your sample.
Catalog No. 50-196-1371