Learn More
Sensorex PH ELECTRODE BNC
Catalog No. NC9660969
Encompass_Preferred
Combination pH/Reference Electrodes with EPOXY BODIES,Standard Laboratory use,GLASS TYPE:Bulb,JUNCTION Pelon,unbreakable electrode is rugged enough to use as a stirring rod,SIZE:DIA :1/4 in. (6mm);LENGTH :6 in. (150mm)
Catalog No. NC9660969 Supplier Sensorex Supplier No. S900C
Combination pH/Reference Electrodes with EPOXY BODIES,Standard Laboratory use,GLASS TYPE:Bulb,JUNCTION Pelon,unbreakable electrode is rugged enough to use as a stirring rod,SIZE:DIA :1/4 in. (6mm);LENGTH :6 in. (150mm)