Learn More
Grainger pH Electrode CPVC BNR Flat Tip
Catalog No. 502768217
Encompass_Preferred
The PH Electrode CPVC BNR Flat Tip is a pH electrode with a flat tip made of CPVC plastic. It has a BNC connector and a 1 meter cable. The electrode has a pH range of 0-14 pH with an accuracy of ±0.1 pH. It has a temperature range of 0-80°C. The flat tip provides fast, accurate pH measurements. The electrode is refillable and requires pH 4 & pH 7 buffers for calibration. It can be used for testing pH in liquids. The electrode is suitable for laboratory, industrial and educational use.
Catalog No. 50-276-8217 Supplier Grainger Supplier No. 16Z016
The PH Electrode CPVC BNR Flat Tip is a pH electrode with a flat tip made of CPVC plastic. It has a BNC connector and a 1 meter cable. The electrode has a pH range of 0-14 pH with an accuracy of ±0.1 pH. It has a temperature range of 0-80°C. The flat tip provides fast, accurate pH measurements. The electrode is refillable and requires pH 4 & pH 7 buffers for calibration. It can be used for testing pH in liquids. The electrode is suitable for laboratory, industrial and educational use.