Learn More
Grainger pH Electrode CPVC BNR Flat Tip
Catalog No. 502768061
Encompass_Preferred
The SensoRex Insertion pH Electrode from Sensorex is a pH electrode designed for insertion into pipelines, tanks, and vessels to measure pH in process applications. It has a 12mm diameter pH sensor with a flat tip and features an epoxy body and PVDF junction for chemical resistance. The electrode has a built-in temperature sensor and 3/4" NPT process connection. It has a measurement range of 0-14 pH with accuracy of ±0.1 pH. The electrode requires a preamplifier sold separately.
Catalog No. 50-276-8061 Supplier Grainger Supplier No. 16Z019
The SensoRex Insertion pH Electrode from Sensorex is a pH electrode designed for insertion into pipelines, tanks, and vessels to measure pH in process applications. It has a 12mm diameter pH sensor with a flat tip and features an epoxy body and PVDF junction for chemical resistance. The electrode has a built-in temperature sensor and 3/4" NPT process connection. It has a measurement range of 0-14 pH with accuracy of ±0.1 pH. The electrode requires a preamplifier sold separately.