Learn More
Grainger pH Electrode Epoxy BNC/Dual banana plug
Catalog No. 502768156
Encompass_Preferred
The SENSOREX S222CD pH Electrode is a glass body combination pH electrode with a double junction reference, ideal for laboratory use. It has a pH range of 0-14 pH, a temperature range of 0-80°C, and 12mm diameter x 120mm length. The S222CD has a BNC connector and 1m cable length. It is made from lead-free glass and is refillable. This electrode is designed for precise pH measurement in aqueous solutions.
Catalog No. 50-276-8156 Supplier Grainger Supplier No. 16Y994
The SENSOREX S222CD pH Electrode is a glass body combination pH electrode with a double junction reference, ideal for laboratory use. It has a pH range of 0-14 pH, a temperature range of 0-80°C, and 12mm diameter x 120mm length. The S222CD has a BNC connector and 1m cable length. It is made from lead-free glass and is refillable. This electrode is designed for precise pH measurement in aqueous solutions.