Learn More
Grainger pH Electrode Epoxy BNC Flat Tip 15mm Dia
Catalog No. 502767957
Encompass_Preferred
The Grainger 16Y970 is a 3M Scotch-Brite General Purpose Hand Pad that features an open web construction with non-woven fibers for general cleaning. It has a 6 inch by 9 inch size, medium duty abrasiveness, and can be used for a variety of cleaning applications including prep work, finish work, and cleaning. The pad works on multiple surfaces including stainless steel, ceramic, plastic and glass. It effectively removes grease, oil, dirt, food residue and more. The 3M Scotch-Brite General Purpose Hand Pad provides versatile cleaning performance.
Catalog No. 50-276-7957 Supplier Grainger Supplier No. 16Y970
The Grainger 16Y970 is a 3M Scotch-Brite General Purpose Hand Pad that features an open web construction with non-woven fibers for general cleaning. It has a 6 inch by 9 inch size, medium duty abrasiveness, and can be used for a variety of cleaning applications including prep work, finish work, and cleaning. The pad works on multiple surfaces including stainless steel, ceramic, plastic and glass. It effectively removes grease, oil, dirt, food residue and more. The 3M Scotch-Brite General Purpose Hand Pad provides versatile cleaning performance.