Learn More
Grainger pH Electrode Epoxy BNC Flat Tip 12mm Dia
Catalog No. 502765149
Encompass_Preferred
The Grainger Approved pH Electrode is a glass body combination electrode designed for general purpose pH measurements. It has a 12 mm diameter pH bulb and gel filled outer body. The electrode has a BNC connector and 1 m cable. It has a 3.5M KCl electrolyte solution. The electrode measures pH 0-14 and has a temperature range of 0-80°C. It has a response time of 95% in 1 minute.
Catalog No. 50-276-5149 Supplier Grainger Supplier No. 16Y986
The Grainger Approved pH Electrode is a glass body combination electrode designed for general purpose pH measurements. It has a 12 mm diameter pH bulb and gel filled outer body. The electrode has a BNC connector and 1 m cable. It has a 3.5M KCl electrolyte solution. The electrode measures pH 0-14 and has a temperature range of 0-80°C. It has a response time of 95% in 1 minute.