Learn More
Grainger pH Electrode Epoxy BNC Round Tip
Catalog No. 502768161
Encompass_Preferred
The Grainger Approved pH Electrode is a combination electrode designed for general purpose pH measurement in the 0-14 pH range. It has a glass body, gel-filled reference with Ag/AgCl reference wire, BNC connector, and epoxy body. The electrode has a response time of less than 10 seconds and is compatible with pH meters that accept BNC connectors. It requires storage solution and regular calibration for accurate pH readings. The electrode measures 12" length x 0.5" diameter.
Catalog No. 50-276-8161 Supplier Grainger Supplier No. 16Y953
The Grainger Approved pH Electrode is a combination electrode designed for general purpose pH measurement in the 0-14 pH range. It has a glass body, gel-filled reference with Ag/AgCl reference wire, BNC connector, and epoxy body. The electrode has a response time of less than 10 seconds and is compatible with pH meters that accept BNC connectors. It requires storage solution and regular calibration for accurate pH readings. The electrode measures 12" length x 0.5" diameter.