Learn More
Grainger pH Electrode Epoxy BNC Round Tip
Catalog No. 502768163
Encompass_Preferred
The Grainger Approved pH Electrode is a combination electrode designed for general purpose pH measurement. It has a glass body, epoxy body material, gel electrolyte, and a BNC connector. The electrode has a pH range of 0-14 pH, is compatible with aqueous solutions, and has a temperature range of 0-80°C. The electrode has a 12 mm diameter x 120 mm length. It should be stored in pH 4 buffer solution.
Catalog No. 50-276-8163 Supplier Grainger Supplier No. 16Y975
The Grainger Approved pH Electrode is a combination electrode designed for general purpose pH measurement. It has a glass body, epoxy body material, gel electrolyte, and a BNC connector. The electrode has a pH range of 0-14 pH, is compatible with aqueous solutions, and has a temperature range of 0-80°C. The electrode has a 12 mm diameter x 120 mm length. It should be stored in pH 4 buffer solution.