Learn More
Grainger pH Electrode Epoxy BNC Round Tip
Catalog No. 502768218
Encompass_Preferred
The Grainger Approved pH Electrode is a glass body combination electrode with a single junction reference, ideal for laboratory use. It has a pH range of 0-14 pH with an accuracy of ±0.02 pH. The body dimensions are 12mm x 150mm with a BNC connector and 1m cable. It should be stored in storage solution when not in use.
Catalog No. 50-276-8218 Supplier Grainger Supplier No. 16Y990
The Grainger Approved pH Electrode is a glass body combination electrode with a single junction reference, ideal for laboratory use. It has a pH range of 0-14 pH with an accuracy of ±0.02 pH. The body dimensions are 12mm x 150mm with a BNC connector and 1m cable. It should be stored in storage solution when not in use.