Learn More
Grainger pH Electrode Epoxy BNC Round Tip
Catalog No. 502768065
Encompass_Preferred
The Grainger Approved pH Electrode Epoxy BNC is a pH electrode with an epoxy body and BNC connector. It has a glass bulb sensing element and measures pH from 0-14 pH with an accuracy of ±0.1 pH. The electrode has a temperature range of 0-80°C. It has a response time of less than 10 seconds and an impedance of less than 500 MΩ. The electrode is 12 mm diameter x 150 mm length. It requires a BNC connector and is compatible with standard pH meters. The electrode is maintenance free with no refilling required.
Catalog No. 50-276-8065 Supplier Grainger Supplier No. 16Y985
The Grainger Approved pH Electrode Epoxy BNC is a pH electrode with an epoxy body and BNC connector. It has a glass bulb sensing element and measures pH from 0-14 pH with an accuracy of ±0.1 pH. The electrode has a temperature range of 0-80°C. It has a response time of less than 10 seconds and an impedance of less than 500 MΩ. The electrode is 12 mm diameter x 150 mm length. It requires a BNC connector and is compatible with standard pH meters. The electrode is maintenance free with no refilling required.