Learn More
Grainger pH Electrode Epoxy BNC Spear Tip
Catalog No. 502767498
Encompass_Preferred
The SensoRex Electrode pH Electrode is a glass body pH electrode with a double junction reference system. It has a pH range of 0-14 pH, a temperature range of 0-80°C, and an impedance of less than 300 MΩ. The electrode has a 12 mm diameter glass body, 1 m cable length, and BNC connector. It is compatible with most pH meters and ideal for laboratory and industrial pH measurement applications. The electrode requires storage solution and regular calibration for accurate pH readings.
Catalog No. 50-276-7498 Supplier Grainger Supplier No. 16Y954
The SensoRex Electrode pH Electrode is a glass body pH electrode with a double junction reference system. It has a pH range of 0-14 pH, a temperature range of 0-80°C, and an impedance of less than 300 MΩ. The electrode has a 12 mm diameter glass body, 1 m cable length, and BNC connector. It is compatible with most pH meters and ideal for laboratory and industrial pH measurement applications. The electrode requires storage solution and regular calibration for accurate pH readings.