Learn More
Grainger pH Electrode Epoxy BNR Round Tip
Catalog No. 502768175
Encompass_Preferred
The SENSOREX S222CD pH Electrode is a glass body combination pH electrode with a double junction reference, ideal for laboratory use. It has a pH range of 0-14 pH, a temperature range of 0-80°C, and 12/120 mm dimensions. The S222CD features a spherical, anti-clogging pH bulb and quick disconnect DIN connector. It has a fast response time and is compatible with standard pH meters. The electrode requires storage solution and regular calibration for optimal performance.
Catalog No. 50-276-8175 Supplier Grainger Supplier No. 16Y962
The SENSOREX S222CD pH Electrode is a glass body combination pH electrode with a double junction reference, ideal for laboratory use. It has a pH range of 0-14 pH, a temperature range of 0-80°C, and 12/120 mm dimensions. The S222CD features a spherical, anti-clogging pH bulb and quick disconnect DIN connector. It has a fast response time and is compatible with standard pH meters. The electrode requires storage solution and regular calibration for optimal performance.